Learn More
Abcam STING Inhibitor, C-176, 25MG
Catalog No. 502671238
Encompass_Preferred
MW 358.09 Da, Purity >=98%. STING inhibitor C-176 is a potent, small-molecule inhibitor of STING (stimulator of interferon genes) protein, that attenuates STING-associated autoinflammatory disease in mice. STING protein is a central signaling component of the intracellular DNA sensing pathway.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1238 Supplier Abcam Supplier No. AB28543825MG
MW 358.09 Da, Purity >=98%. STING inhibitor C-176 is a potent, small-molecule inhibitor of STING (stimulator of interferon genes) protein, that attenuates STING-associated autoinflammatory disease in mice. STING protein is a central signaling component of the intracellular DNA sensing pathway.
The product is subject to the following: Abcam Restricted Use Statement