Learn More
Selleck Chemical LLC STING inhibitor C-178 2mg
SDP
Catalog No. 502173202
Encompass_Preferred
C-178 is a covalent inhibitor of STING,covalently bind to Cys91. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3202 Supplier Selleck Chemical LLC Supplier No. S66672MG
C-178 is a covalent inhibitor of STING,covalently bind to Cys91. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.