Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STING ligand-1 | 2365039-41-2 | 99.7% | 523.98 | 5 MG
SDP

Catalog No. 5000364616
Encompass_Preferred

STING ligand-1 is a lead STING ligand with an IC50 of 68 nM for HAQ STING. It exhibits functional inhibition of STING mediated cytokine release.

  • Appearance: Solid
  • Color: Off-white to light yellow
  • Purity: 99.65%
  • CAS number: 2365039-41-2
  • Molecular weight: 523.98
  • Formula: C29H27ClFNO5
  • Shipping: Room temperature in continental US
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month
  • IC50: 68 nM (HAQ STING)

Catalog No. 50-003-64616 Supplier Medchemexpress LLC Supplier No. 5MGHY114399
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STING ligand-1 is a lead STING ligand with an IC50 of 68 nM for HAQ STING. It exhibits functional inhibition of STING mediated cytokine release.

  • Appearance: Solid
  • Color: Off-white to light yellow
  • Purity: 99.65%
  • CAS number: 2365039-41-2
  • Molecular weight: 523.98
  • Formula: C29H27ClFNO5
  • Shipping: Room temperature in continental US
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month
  • IC50: 68 nM (HAQ STING)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.