Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STING ligand-3 | 2762552-73-6 | 99.2% | C15H11N3O7 | 5 MG
SDP

Catalog No. 5000358281
Encompass_Preferred

STING ligand-3 is supplied as a light yellow to yellow solid. This product has been rigorously tested and complies with the given specifications, exhibiting a high purity of 99.2%.

  • Supplied as a light yellow to yellow solid
  • High purity of 99.2% by HPLC
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Chemical formula: C15H11N3O7

Catalog No. 50-003-58281 Supplier Medchemexpress LLC Supplier No. HY477095MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STING ligand-3 is supplied as a light yellow to yellow solid. This product has been rigorously tested and complies with the given specifications, exhibiting a high purity of 99.2%.

  • Supplied as a light yellow to yellow solid
  • High purity of 99.2% by HPLC
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
  • Chemical formula: C15H11N3O7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.