Learn More
MSC STINGER BERM
Catalog No. 5000687462
Encompass
The TPU 3010 is an environmental thermoplastic material designed for secondary containment applications. It features a robust polyester fabric with a PVC-PU blend coating.
  • Polyester 1100 Dtex fabric
  • PVC-PU blend coating
  • Weighs 1015 gr/m2
  • Ideal for secondary containment
Catalog No. 50-006-87462 Supplier MSC Supplier No. 07572415
The TPU 3010 is an environmental thermoplastic material designed for secondary containment applications. It features a robust polyester fabric with a PVC-PU blend coating.
  • Polyester 1100 Dtex fabric
  • PVC-PU blend coating
  • Weighs 1015 gr/m2
  • Ideal for secondary containment