Learn More
Aviva Systems Biology Corp STIP1 Antibody - N-terminal region (ARP46164_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46164_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against STIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STIP1. Alternate names: HOP, IEF-SSP-3521, P60, STI1, STI1LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-59256