Learn More
Chemglass Life Sciences Chemglass Stir Bar, PTFE-Coated Magnetic Micro, 10 x 3 mm, Pack of 50 - for magnetic stirrers
SDP
Catalog No. NC2022812
Encompass_Preferred
Chemglass PTFE-coated magnetic micro stir bars provide reliable mixing in very small volumes. Each bar measures 10 x 3 mm and is bulk packed in a 50-piece set for convenient laboratory use. The PTFE coating meets FDA-grade and USP Class VI standards for chemical resistance and biocompatibility.
  • PTFE coating with FDA-grade and USP Class VI compliance.
  • Micro size: 10 x 3 mm.
  • Bulk pack: 50 pieces per pack.
  • Designed for use with magnetic stirrers and hot plate stirrers.
  • Suitable for small-volume mixing and agitation.
Catalog No. NC2022812 Supplier Chemglass Life Sciences Supplier No. CG2003160
Chemglass PTFE-coated magnetic micro stir bars provide reliable mixing in very small volumes. Each bar measures 10 x 3 mm and is bulk packed in a 50-piece set for convenient laboratory use. The PTFE coating meets FDA-grade and USP Class VI standards for chemical resistance and biocompatibility.
  • PTFE coating with FDA-grade and USP Class VI compliance.
  • Micro size: 10 x 3 mm.
  • Bulk pack: 50 pieces per pack.
  • Designed for use with magnetic stirrers and hot plate stirrers.
  • Suitable for small-volume mixing and agitation.