Learn More
Cole-Parmer Mono-Mold Slim-Line Stir Bar with Spinning Ring, 2"L x 5/16" dia
Catalog No. NC0484864 Shop All Cole-Parmer Products
Encompass_Preferred
Slim-Line stir bars feature a slimmer profile and larger length to diameter ratio. Available with or without spinning rings.
Catalog No. NC0484864 Supplier Cole-Parmer Supplier No. 0477565
Slim-Line stir bars feature a slimmer profile and larger length to diameter ratio. Available with or without spinning rings.