Learn More
Chemglass Life Sciences Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 5 x 2, Bulk Packed
SDP
Catalog No. 501531393
Encompass_Preferred
Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 5 x 2, Bulk Packed. Micro PTFE stir bar for the very smallest containers. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. 50-153-1393 Supplier Chemglass Life Sciences Supplier No. CG2003120
Stir Bar, Micro, Magnetic, PTFE, Length x Dia. in mm: 5 x 2, Bulk Packed. Micro PTFE stir bar for the very smallest containers. Manufactured with FDA-grade and USP Class VI PTFE.