Learn More
Hach Company Stir Bar, 8 x 3 mm - replacement for 5500sc Ammonia Monochloramine Analyzer
Catalog No. NC2398715 Shop All Hach Company Products
Encompass
Replacement magnetic stir bar for use with the 5500sc Ammonia Monochloramine Analyzer. It provides consistent mixing in the sample chamber to support accurate, continuous measurements; regular replacement is recommended to maintain analyzer performance.
  • Dimensions: 8 x 3 mm.
  • Suggested replacement schedule: 6 months.
  • Material: not specified by the manufacturer.
  • Function: maintains proper mixing in the sample chamber.
Catalog No. NC2398715 Supplier Hach Company Supplier No. 9563900
Replacement magnetic stir bar for use with the 5500sc Ammonia Monochloramine Analyzer. It provides consistent mixing in the sample chamber to support accurate, continuous measurements; regular replacement is recommended to maintain analyzer performance.
  • Dimensions: 8 x 3 mm.
  • Suggested replacement schedule: 6 months.
  • Material: not specified by the manufacturer.
  • Function: maintains proper mixing in the sample chamber.