Learn More
Hach Company Stir Bar Cl17 before 3-2020
Supplier:  Hach Company 4425700
Encompass
Stir Bar for Cl17 (Prior to March 2000)The stir bar for the Cl17 (produced prior to March 2000) provides mixing for the sample and the instrument s reagents Stir bar size is 5/16in x 3/8in
Catalog No. 50-002-07188