Learn More
Hach Company Stir Bar, Cuvette
Supplier:  Hach Company 2140800
Encompass
PTFE magnetic centrifugal stirrer fits into standard 10 mm light path spectral cell and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirrer. This one-piece molded unit will not scratch cuvettes. May also be used in test tubes.
Catalog No. 50-199-5468