Learn More
Thomas Scientific Stir Bar, Cylindrical, PTFE, 15mm, Bag of 2
SDP
Catalog No. 501457333
Encompass
Produced from PTFE For the stirring of liquids on magnetic stirrers and/or hotplate stirrers Strong alloy magnets sheathed in pure white inert PTFE Supplied in polarity aligned packs to ensure maximum coupling strength Resistant to virtually all chemical attack and temperatures up to 260°C
Catalog No. 50-145-7333 Supplier Thomas Scientific Supplier No. 1181B91
Produced from PTFE For the stirring of liquids on magnetic stirrers and/or hotplate stirrers Strong alloy magnets sheathed in pure white inert PTFE Supplied in polarity aligned packs to ensure maximum coupling strength Resistant to virtually all chemical attack and temperatures up to 260°C