Learn More
Thomas Scientific Stir Bar, Cylindrical, PTFE, 25mm, Bag of 2
SDP
Catalog No. 501457334
Encompass
Produced from PTFE For the stirring of liquids on magnetic stirrers and/or hotplate stirrers Strong alloy magnets sheathed in pure white inert PTFE Supplied in polarity aligned packs to ensure maximum coupling strength Resistant to virtually all chemical attack and temperatures up to 260°C
Catalog No. 50-145-7334 Supplier Thomas Scientific Supplier No. 1181B95
Produced from PTFE For the stirring of liquids on magnetic stirrers and/or hotplate stirrers Strong alloy magnets sheathed in pure white inert PTFE Supplied in polarity aligned packs to ensure maximum coupling strength Resistant to virtually all chemical attack and temperatures up to 260°C