Learn More
Hach Company Stir Bar, Magnetic Egg-Shaped
Supplier:  Hach Company 2753200
Encompass
Designed specifically for round bottom flasks and vessels. The PTFE coated, egg-shaped, magnetic stir bar fits the round bottom for uniform stirring without restriction. Recommended for the following round bottom flask sizes: 10, 25, 50, and 100 mL.
Catalog No. 50-199-6623