Learn More
Cole-Parmer Egg-Shaped Stir Bar, 13/16" L x 3/8" Dia; 1/Pk
Supplier:  Cole-Parmer 0476820
Encompass_Preferred
Egg-shaped stir bars have oval magnets—ideal for use in round-bottom flasks. Shape conforms to flask bottom and ensures complete mixing.
Catalog No. NC0553423