Learn More
V&P Scientific Inc STIR ELEMENT, CYLINDER, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, Used in Bottles, Microplates, Tubes/Vials, 100/pack
Supplier:  V&P Scientific Inc VP 77512
Encompass
STIR ELEMENT, CYLINDER, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, 3.0mm Diameter, 12.0mm Length, 260°C Max Operating Temp for Coating
Catalog No. NC1546700