Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Triangular, PTFE, 12mm L X 6mm Diameter
SDP
Catalog No. 501531395
Encompass_Preferred
Stir Bar, Magnetic, Triangular, PTFE, 12mm L X 6mm Diameter. Effective for dissolving solids and mixing sediments in a flat bottom container such as a beaker or erlenmeyer flask. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. 50-153-1395 Supplier Chemglass Life Sciences Supplier No. CG200910
Stir Bar, Magnetic, Triangular, PTFE, 12mm L X 6mm Diameter. Effective for dissolving solids and mixing sediments in a flat bottom container such as a beaker or erlenmeyer flask. Manufactured with FDA-grade and USP Class VI PTFE.