Learn More
Chemglass Life Sciences Chemglass Stir Bar, Magnetic, PTFE, Turbo Rare-Earth Elliptical, 10 x 6 mm, for magnetic stirrers
SDP
Catalog No. NC1810970
Encompass_Preferred
Chemglass CG-2005T-01 is a PTFE-coated turbo (elliptical) magnetic stir bar with a rare-earth magnet. It measures 10 x 6 mm and is designed for use with laboratory magnetic stirrers for mixing and agitation.
  • Dimensions: 10 x 6 mm.
  • Material: PTFE.
  • Shape: Elliptical (turbo).
  • Magnet type: Rare-earth magnet.
  • Pack quantity: Each.
  • Intended use: For magnetic stirring and laboratory mixing.
Catalog No. NC1810970 Supplier Chemglass Life Sciences Supplier No. CG2005T01
Chemglass CG-2005T-01 is a PTFE-coated turbo (elliptical) magnetic stir bar with a rare-earth magnet. It measures 10 x 6 mm and is designed for use with laboratory magnetic stirrers for mixing and agitation.
  • Dimensions: 10 x 6 mm.
  • Material: PTFE.
  • Shape: Elliptical (turbo).
  • Magnet type: Rare-earth magnet.
  • Pack quantity: Each.
  • Intended use: For magnetic stirring and laboratory mixing.