Learn More
Hach Company Stir Bar, Magnetic, Plus Shape, 3/8"
Supplier:  Hach Company 2928601
Encompass
This magnetic stir bar adds speed and efficiency to your mixing requirements. The “+” shape creates a deep vortex and provides stable, quiet operation.
Catalog No. 50-199-6818