Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Triangular, PTFE, 35mm L X 9mm Diameter
SDP
Catalog No. 50872478
Encompass_Preferred
Stir Bar, Magnetic, Triangular, PTFE, 35mm L X 9mm Diameter. Effective for dissolving solids and mixing sediments in a flat bottom container such as a beaker or Erlenmeyer flask. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. 50-872-478 Supplier Chemglass Life Sciences Supplier No. CG200914
Stir Bar, Magnetic, Triangular, PTFE, 35mm L X 9mm Diameter. Effective for dissolving solids and mixing sediments in a flat bottom container such as a beaker or Erlenmeyer flask. Manufactured with FDA-grade and USP Class VI PTFE.