Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Triangular, PTFE, 50mm L X 12mm Diameter
SDP
Catalog No. 50872480
Encompass_Preferred
Stir Bar, Magnetic, Triangular, PTFE, 50mm L X 12mm Diameter. Effective for dissolving solids and mixing sediments in a flat bottom container such as a beaker or Erlenmeyer flask. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. 50-872-480 Supplier Chemglass Life Sciences Supplier No. CG200916
Stir Bar, Magnetic, Triangular, PTFE, 50mm L X 12mm Diameter. Effective for dissolving solids and mixing sediments in a flat bottom container such as a beaker or Erlenmeyer flask. Manufactured with FDA-grade and USP Class VI PTFE.