Learn More
Chemglass Life Sciences Stir Bar, Magnetic, "X" Shaped, PTFE, 38 x 11: Diameter x Height (mm)
SDP
Catalog No. 50872487
Encompass_Preferred
Stir Bar, Magnetic, "X" Shaped, PTFE, 38 x 11: Diameter x Height (mm). Shape of stir bar creates a deep vortex and provides stability during stirring. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. 50-872-487 Supplier Chemglass Life Sciences Supplier No. CG201305
Stir Bar, Magnetic, "X" Shaped, PTFE, 38 x 11: Diameter x Height (mm). Shape of stir bar creates a deep vortex and provides stability during stirring. Manufactured with FDA-grade and USP Class VI PTFE.