Learn More
Chemglass Life Sciences Stir Bar, Magnetic, "X" Shaped, PTFE, 20 x 8: Diameter x Height (mm)
SDP
Catalog No. 50872484
Encompass_Preferred
Stir Bar, Magnetic, "X" Shaped, PTFE, 20 x 8: Diameter x Height (mm). Shape of stir bar creates a deep vortex and provides stability during stirring. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. 50-872-484 Supplier Chemglass Life Sciences Supplier No. CG201302
Stir Bar, Magnetic, "X" Shaped, PTFE, 20 x 8: Diameter x Height (mm). Shape of stir bar creates a deep vortex and provides stability during stirring. Manufactured with FDA-grade and USP Class VI PTFE.