Learn More
Electron Microscopy Sciences Micro Stir Bars, 1.5 x 15 mm
SDP
Catalog No. 501899180
Encompass_Preferred
Coated with "PTFE" white resin. For stirring test tubes, micro-beakers, and more. Packed 12 bars per box. 1/16 x 9/16 inches; 1.5x15mm
Catalog No. 50-189-9180 Supplier Electron Microscopy Sciences Supplier No. 6451501
Coated with "PTFE" white resin. For stirring test tubes, micro-beakers, and more. Packed 12 bars per box. 1/16 x 9/16 inches; 1.5x15mm