Learn More
Hach Company Stir Bar, Micro, 3 x 10 mm
Supplier:  Hach Company 2054959
Encompass
Use micro stir bar for mixing in vials, test tubes, spectrophotometer cells and other small-volume containers. Usable at temperatures up to 260°C. Alnico V magnet encapsulated in PTFE.
Catalog No. 50-199-5186