Learn More
Anton Paar Pack of 10 PTFE coated stir bars (3x10 mm)
Catalog No. NC1818282
Encompass_Preferred
Pack of 10 cylindrical PTFE coated stir bars (3x10 mm) for use with 10 mL Reaction Vial G10, item NC1110206.Suitable for use with Anton Paar Monowave 50
Catalog No. NC1818282 Supplier Anton Paar Supplier No. 89088
Pack of 10 cylindrical PTFE coated stir bars (3x10 mm) for use with 10 mL Reaction Vial G10, item NC1110206.Suitable for use with Anton Paar Monowave 50