Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, Turbo, Rare Earth, Elliptical, 50 x 24mm
SDP
Catalog No. NC1876265
Encompass_Preferred
Exceptional strength coupled with almost total resistance to demagnetisation. Identified by an inert carbon black spot.
Catalog No. NC1876265 Supplier Chemglass Life Sciences Supplier No. CG2005T04
Exceptional strength coupled with almost total resistance to demagnetisation. Identified by an inert carbon black spot.