Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, Turbo, Rare Earth, Elliptical, 70 x 28mm
SDP
Catalog No. NC1876263
Encompass_Preferred
Exceptional strength coupled with almost total resistance to demagnetisation. Identified by an inert carbon black spot.
Catalog No. NC1876263 Supplier Chemglass Life Sciences Supplier No. CG2005T05
Exceptional strength coupled with almost total resistance to demagnetisation. Identified by an inert carbon black spot.