Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- For use with hotplates and stirrers
- Use this device to quickly and easily remove stir bars
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | Hotplate-Stirrer e-G51 HS07 C 120 V US, Hotplate-Stirrer e-G51 HSRDM 120 V US, Hotplate-Stirrer e-G51 HS10 C 120 V US, Stirrer e-G51 ST07 C 120 V US, Hotplate-Stirrer e-G71 HSRDM 120 V US, Hotplate-Stirrer e-G71 HS10 C 120 V US, Hotplate-Stirrer e-G71 HS07 C 120 V US |
| Type | Stir Bar Retriever |
| Material | Metal |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
