Learn More
Wilmad Labglass STIR BEARING COMP 24/40
Supplier:  Wilmad Labglass LG9455100
Encompass
Stirrer Bearing, PTFE Sealed, [ST] Joint. Joint Size: [ST] 24/40 Inner. Design allows liquid or vapors to contact glass and PTFE only. PTFE sleeve with O-Ring is bored for use with 10mm OD polished Stirrer shaft.
Catalog No. 50-805-560