Learn More
Wilmad Labglass STIR BLADE PTFE 1/8IN 24X110MM
Supplier:  Wilmad Labglass LG9510T106
Encompass
Stirrer Blade, PTFE. For Shaft OD: 10mm; Blade Length x Width: 24mm x 110mm; Fits Flask Capacity: 2000mL to 3000mL; Minimum [ST] Joint Clearance: 29/42. Made of 1/8 in. polytetrafluoroethylene (PTFE)
Catalog No. 50-807-695