Learn More
Ace Glass, Inc. Stir blade, stainless, 24x80mm, holed, 10mm shaft
SDP
Catalog No. 50979677
Encompass_Preferred
316 stainless steel, football shaped, holed stir blades for 10mm OD button type stir shafts.
Catalog No. 50-979-677 Supplier Ace Glass, Inc. Supplier No. 809672
316 stainless steel, football shaped, holed stir blades for 10mm OD button type stir shafts.