Learn More
Ace Glass, Inc. Stir blade, stainless, 24x160mm, 10mm shaft
SDP
Catalog No. NC0751863
Encompass_Preferred
24X160mm 304 SS stir blades for 10mm O.D. button type stir shafts. For compatible stir shafts see ACE 8068, 8070, 8074, and 8075 series.
Catalog No. NC0751863 Supplier Ace Glass, Inc. Supplier No. 808616
24X160mm 304 SS stir blades for 10mm O.D. button type stir shafts. For compatible stir shafts see ACE 8068, 8070, 8074, and 8075 series.