Learn More
Chemglass Life Sciences Stir Bar Retriever, PTFE, 450 x 10 mm - For magnetic stir bars
SDP
Catalog No. NC3238461
Encompass_Preferred
Designed to recover magnetic stir bars from deep reaction vessels, this PTFE-coated stir bar retriever measures 450 x 10 mm and provides chemically inert, autoclavable performance. Its long shaft and magnetic tip allow safe, convenient retrieval without contaminating samples.
  • Inert PTFE construction for chemical resistance.
  • 450 x 10 mm length and diameter for deep vessels.
  • Autoclavable for laboratory reuse.
  • Molded in FDA-approved PTFE for leak-free performance.
  • Available with Alnico V or rare earth magnet options.
  • Designed to retrieve magnetic stir bars without sample contamination.
Catalog No. NC3238461 Supplier Chemglass Life Sciences Supplier No. CG2015T04
Designed to recover magnetic stir bars from deep reaction vessels, this PTFE-coated stir bar retriever measures 450 x 10 mm and provides chemically inert, autoclavable performance. Its long shaft and magnetic tip allow safe, convenient retrieval without contaminating samples.
  • Inert PTFE construction for chemical resistance.
  • 450 x 10 mm length and diameter for deep vessels.
  • Autoclavable for laboratory reuse.
  • Molded in FDA-approved PTFE for leak-free performance.
  • Available with Alnico V or rare earth magnet options.
  • Designed to retrieve magnetic stir bars without sample contamination.