Learn More
Wilmad Labglass STIR BRNG 10MM PTFE COMP 24/40
Catalog No. 50806493
Encompass_Preferred
Stirrer Bearing, Precision, PTFE, Complete, 10mm. Joint Size: [ST] 24/40. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.
Catalog No. 50-806-493 Supplier Wilmad Labglass Supplier No. LG9444100
Stirrer Bearing, Precision, PTFE, Complete, 10mm. Joint Size: [ST] 24/40. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.