Learn More
Wilmad Labglass STIR BRNG 10MM PTFE COMP 29/42
Catalog No. 50804341
Encompass_Preferred
Stirrer Bearing, Precision, PTFE, Complete, 10mm. Joint Size: [ST] 29/42. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.
Catalog No. 50-804-341 Supplier Wilmad Labglass Supplier No. LG9444104
Stirrer Bearing, Precision, PTFE, Complete, 10mm. Joint Size: [ST] 29/42. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.