Learn More
Wilmad Labglass STIR BRNG 10MM PTFE COMP 34/45
Catalog No. 50803749
Encompass_Preferred
Stirrer Bearing, Precision, PTFE, Complete, 10mm. Joint Size: [ST] 34/45. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.
Catalog No. 50-803-749 Supplier Wilmad Labglass Supplier No. LG9444106
Stirrer Bearing, Precision, PTFE, Complete, 10mm. Joint Size: [ST] 34/45. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.