Learn More
Wilmad Labglass STIR BRNG 10MM PTFE COMP 45/50
Catalog No. 50806577
Encompass_Preferred
Stirrer Bearing, Precision, PTFE, Complete, 10mm. Joint Size: [ST] 45/50. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.
Catalog No. 50-806-577 Supplier Wilmad Labglass Supplier No. LG9444108
Stirrer Bearing, Precision, PTFE, Complete, 10mm. Joint Size: [ST] 45/50. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.