Learn More
Wilmad Labglass STIR BRNG PTFE COMP 55/50
Supplier:  Wilmad Labglass LG9450102
Encompass
Stirrer Bearing, Precision, PTFE, Complete, 19mm. Joint Size: [ST] 55/50. Complete assembly includes PTFE Stirrer Bearing, PTFE bushing, saddle O-Ring, threaded heavy wall glass adapter and top screw cap.
Catalog No. 50-806-230