Learn More
V&P Scientific Inc STIR ELEMENT, CROSS, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, Used in Tubes/Vials, 10/pack
Catalog No. NC1920808
Encompass
STIR ELEMENT, CROSS, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, 19.0mm Diameter, 8.0mm Height, 260°C Max Operating Temp for Coating
Catalog No. NC1920808 Supplier V&P Scientific Inc Supplier No. VP 77319
STIR ELEMENT, CROSS, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, 19.0mm Diameter, 8.0mm Height, 260°C Max Operating Temp for Coating