Learn More
Azzota Corp Stir Heating Mantle 250ml 250W
Catalog No. 5000887318
Encompass_Preferred
Digital Stirring Heating Mantle Capacity 250ml 250W Maximum temperature is 450 C Stir speed 0 to 1400rpm
Catalog No. 50-008-87318 Supplier Azzota Corp Supplier No. DSHM250
Digital Stirring Heating Mantle Capacity 250ml 250W Maximum temperature is 450 C Stir speed 0 to 1400rpm