Learn More
Yamato Scientific America, Inc. Yamato Magnetic Stirrer with Hot Plate, Max hot plate temp 250°C. Stirring rate up to 1500 rpm, 115V/220V.
Catalog No. 501898191 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Six point controllable magnetic stirrer with chemical resistant ceramic coating hot plate. Rotation and heating can be adjusted individually. Equipped with circuit protector. Stirring rate 300~1500rpm. Stirring capacity 100~2000 ml x 6 pcs. and maximum hot plate temp of 250°C.
Catalog No. 50-189-8191 Supplier Yamato Scientific America, Inc. Supplier No. MG600H115V
Six point controllable magnetic stirrer with chemical resistant ceramic coating hot plate. Rotation and heating can be adjusted individually. Equipped with circuit protector. Stirring rate 300~1500rpm. Stirring capacity 100~2000 ml x 6 pcs. and maximum hot plate temp of 250°C.