Learn More
Ace Glass, Inc. 1oz Stir-Lube stirrer lubrication
SDP
Catalog No. 509558285
Encompass_Preferred
1oz Stir-Lube stirrer lubricant is a superior, low melting, silicone based lubricant which liquefies at body temperature. Its composition allows for application of a very thin layer to a stirring shaft to increase bearing and shaft life at least three times over that of a bearing lubricated with glycerine.
Catalog No. 50-955-8285 Supplier Ace Glass, Inc. Supplier No. 811710
1oz Stir-Lube stirrer lubricant is a superior, low melting, silicone based lubricant which liquefies at body temperature. Its composition allows for application of a very thin layer to a stirring shaft to increase bearing and shaft life at least three times over that of a bearing lubricated with glycerine.