Learn More
Azzota Corp Stir Mantle 1000ml 716F 220V
Catalog No. 5000887267
Encompass_Preferred
Stirring Heating Mantle Capacity 1000ml Maximum temperature is 716 F (380 C) Stir speed 0 to 1400rpm 220V
Catalog No. 50-008-87267 Supplier Azzota Corp Supplier No. SHM1000220V
Stirring Heating Mantle Capacity 1000ml Maximum temperature is 716 F (380 C) Stir speed 0 to 1400rpm 220V