Learn More
Azzota Corp Stir Mantle 3000ml 716F 220V
Catalog No. 5000887302
Encompass_Preferred
Stirring Heating Mantle Capacity 3000ml Maximum temperature is 716 F (380 C) Stir speed 0 to 1400rpm 220V
Catalog No. 50-008-87302 Supplier Azzota Corp Supplier No. SHM3000220V
Stirring Heating Mantle Capacity 3000ml Maximum temperature is 716 F (380 C) Stir speed 0 to 1400rpm 220V