Learn More
Chemglass Life Sciences Agitator, PTFE, 10mm, Four Blade, 50mm Dia, Fits Shaft Size 10mm, 45° Impeller Angle
SDP
Catalog No. CG209101
Encompass_Preferred
Agitator, PTFE, 10mm, Four Blade, 50mm Dia, Fits Shaft Size 10mm, 45¿ Impeller Angle. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 10mm OD stirrer shaft CG-2075-B. Available in 45¿, 60¿ and 90¿ impeller angles. The 45¿ and 60¿ impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. Agitator hub is open so that it can be positioned anywhere along the stirrer shaft and locks onto shaft with a glass filled TFE screw.
Catalog No. CG209101 Supplier Chemglass Life Sciences Supplier No. CG209101
Agitator, PTFE, 10mm, Four Blade, 50mm Dia, Fits Shaft Size 10mm, 45¿ Impeller Angle. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 10mm OD stirrer shaft CG-2075-B. Available in 45¿, 60¿ and 90¿ impeller angles. The 45¿ and 60¿ impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. Agitator hub is open so that it can be positioned anywhere along the stirrer shaft and locks onto shaft with a glass filled TFE screw.