Learn More
Wilmad Labglass STIR PS JKT HD BEARNG 45/50
Catalog No. 50804693
Encompass_Preferred
Stirrer Bearing, Precision, 19mm, Jacketed. Joint Size: [ST] 45/50. Glass sleeve Bearing having a bore of 19mm specially processed for use with LG-9504-series precision ground Stirrer shafts. Bearing is jacketed.
Catalog No. 50-804-693 Supplier Wilmad Labglass Supplier No. LG9427140
Stirrer Bearing, Precision, 19mm, Jacketed. Joint Size: [ST] 45/50. Glass sleeve Bearing having a bore of 19mm specially processed for use with LG-9504-series precision ground Stirrer shafts. Bearing is jacketed.