Learn More
Cole-Parmer Cole-Parmer Polypropylene Stir Rods, 10" x 7mm, 12/pk
Supplier:  Cole-Parmer 0636946
Encompass_Preferred
Stir rods are made of polypropylene and are rounded at both ends. Maximum temperature is 248°F (120°C). Autoclavable.
Catalog No. NC1553806