Learn More
V&P Scientific Inc STIR ELEMENT, CYLINDER, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, Used in Bottles, Microplates, Tubes/Vials, 10/pack
Catalog No. NC1373314
Encompass
STIR ELEMENT, CYLINDER, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, 3.0mm Diameter, 20.0mm Length, 260°C Max Operating Temp for Coating
Catalog No. NC1373314 Supplier V&P Scientific Inc Supplier No. VP 77520
STIR ELEMENT, CYLINDER, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, 3.0mm Diameter, 20.0mm Length, 260°C Max Operating Temp for Coating